Gene Transcripts
- Gene Symbol
- AKR1C6P
- Entrez Gene ID
- 389932
- Gene Name
- aldo-keto reductase family 1 member C6, pseudogene
- Gene Aliases
- TAKR
- Chromosome Location
- Chr. 10: 4871667 - 4916273 on Build GRCh38
- UniGene ID
- Hs.631812
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026743.1 |
|
8 |
833 |
|
GenBank mRNA |
AB037903.1 |
- |
4 |
283 |
|
View Full Product Details