Gene Transcripts

Gene Symbol
C21orf62-AS1
Entrez Gene ID
54067
Gene Name
C21orf62 antisense RNA 1
Gene Aliases
C21orf49
Chromosome Location
Chr. 21: 32772100 - 32797707 on Build GRCh38
UniGene ID
Hs.54725
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024622.1 4 491
NR_024623.1 4 479
GenBank mRNA AK096601.1 - 4 479
BC034794.1 - 2 205
BC117397.1 - 4 373
BC117399.1 - 4 385
DA776905.1 - 4 479


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BX477211.1 - 4 429 Chr. 21 - 32772162 - 32797168 [+]
GenBankmRNA AV717800.1 - 3 283 Chr. 21 - 32772174 - 32797446 [+]

View Full Product Details