Gene Transcripts
- Gene Symbol
- SCARNA9
- Entrez Gene ID
- 619383
- Gene Name
- small Cajal body-specific RNA 9
- Gene Aliases
- Z32, mgU2-19/30
- Chromosome Location
- Chr. 11: 93721514 - 93721866 on Build GRCh38
- UniGene ID
- Hs.660668
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002569.2 |
|
1 |
215 |
|
GenBank mRNA |
AV737883.1 |
- |
1 |
260 |
|
C19000.1 |
- |
1 |
192 |
|
View Full Product Details