Gene Transcripts

Gene Symbol
HCG18
Entrez Gene ID
414777
Gene Name
HLA complex group 18 (non-protein coding)
Gene Aliases
Chromosome Location
Chr. 6: 30287397 - 30327156 on Build GRCh38
UniGene ID
Hs.485041
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024052.2 5 5798
NR_024053.2 4 5584
NR_102326.1 3 7455
NR_102327.1 3 2188
GenBank mRNA AK023334.1 - 1 954
BC040058.1 - 1 843

View Full Product Details