Gene Transcripts
- Gene Symbol
- HCG4
- Entrez Gene ID
- 54435
- Gene Name
- HLA complex group 4 (non-protein coding)
- Gene Aliases
- HCG4P10, HCGIV-10, HCGIV.9
- Chromosome Location
- Chr. 6: 29791031 - 29793073 on Build GRCh38
- UniGene ID
- Hs.660431
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002139.2 |
|
1 |
1996 |
|
GenBank mRNA |
AF036977.1 |
- |
1 |
1994 |
|
BC011879.1 |
- |
1 |
1780 |
|
View Full Product Details