Gene Transcripts
- Gene Symbol
- GUCY2GP
- Entrez Gene ID
- 390003
- Gene Name
- guanylate cyclase 2G, pseudogene
- Gene Aliases
- GUCY2G
- Chromosome Location
- Chr. 10: 112308178 - 112356595 on Build GRCh38
- UniGene ID
- Hs.534691
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028134.1 |
|
6 |
1165 |
|
View Full Product Details