Gene Transcripts
- Gene Symbol
- LINC00469
- Entrez Gene ID
- 283982
- Gene Name
- long intergenic non-protein coding RNA 469
- Gene Aliases
- C17orf54
- Chromosome Location
- Chr. 17: 73749270 - 73828537 on Build GRCh38
- UniGene ID
- Hs.464079
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027146.1 |
|
4 |
899 |
|
GenBank mRNA |
AK097638.1 |
- |
4 |
1017 |
|
View Full Product Details