Gene Transcripts

Gene Symbol
BTN2A3P
Entrez Gene ID
54718
Gene Name
butyrophilin subfamily 2 member A3, pseudogene
Gene Aliases
BTN2.3, BTN2A3
Chromosome Location
Chr. 6: 26421391 - 26430588 on Build GRCh38
UniGene ID
Hs.729655
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027795.1 3 821
GenBank mRNA AK056871.1 - 3 801
AK292658.1 - 3 821
BC143636.1 - 3 456

View Full Product Details