Gene Transcripts
- Gene Symbol
- TSPY26P
- Entrez Gene ID
- 128854
- Gene Name
- testis specific protein, Y-linked 26, pseudogene
- Gene Aliases
- TSPYL3, bA392M18.1
- Chromosome Location
- Chr. 20: 32189146 - 32190360 on Build GRCh38
- UniGene ID
- Hs.647447
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002781.1 |
|
1 |
659 |
|
GenBank mRNA |
BC101556.1 |
- |
1 |
659 |
|
BC101558.1 |
- |
1 |
659 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ432315.1 |
- |
1 |
182 |
|
Chr. 20 - 32189136 - 32189883 [-] |
View Full Product Details