Gene Transcripts
- Gene Symbol
- SNORA11
- Entrez Gene ID
- 677799
- Gene Name
- small nucleolar RNA, H/ACA box 11
- Gene Aliases
- SNORA11A, U107
- Chromosome Location
- Chr. X: 54814370 - 54814500 on Build GRCh38
- UniGene ID
- Hs.679783
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002953.1 |
|
1 |
94 |
|
GenBank mRNA |
AM055729.1 |
- |
1 |
94 |
|
- Gene Symbol
- MAGED2
- Entrez Gene ID
- 10916
- Gene Name
- MAGE family member D2
- Gene Aliases
- 11B6, BARTS5, BCG-1, BCG1, HCA10, MAGE-D2
- Chromosome Location
- Chr. X: 54807599 - 54816015 on Build GRCh38
- UniGene ID
- Hs.522665
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK091003.1 |
- |
1 |
925 |
|
View Full Product Details