Gene Transcripts
- Gene Symbol
- FAM41C
- Entrez Gene ID
- 284593
- Gene Name
- family with sequence similarity 41 member C
- Gene Aliases
- Chromosome Location
- Chr. 1: 868071 - 876802 on Build GRCh38
- UniGene ID
- Hs.449006
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027055.1 |
|
2 |
150 |
|
View Full Product Details