Gene Transcripts
- Gene Symbol
- SNORA29
- Entrez Gene ID
- 677812
- Gene Name
- small nucleolar RNA, H/ACA box 29
- Gene Aliases
- ACA29
- Chromosome Location
- Chr. 6: 159785594 - 159785733 on Build GRCh38
- UniGene ID
- Hs.363137
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002965.1 |
|
1 |
71 |
|
GenBank mRNA |
AJ609472.1 |
- |
1 |
71 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA429995.1 |
- |
1 |
209 |
|
Chr. 6 - 159784784 - 159785871 [-] |
View Full Product Details