Gene Transcripts
- Gene Symbol
- UGT2A3
- Entrez Gene ID
- 79799
- Gene Name
- UDP glucuronosyltransferase family 2 member A3
- Gene Aliases
- Chromosome Location
- Chr. 4: 68928459 - 68951800 on Build GRCh38
- UniGene ID
- Hs.122583
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024010.1 |
|
3 |
907 |
|
GenBank mRNA |
AK293381.1 |
- |
3 |
478 |
|
BP262099.1 |
- |
3 |
205 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BP261788.1 |
- |
3 |
206 |
|
Chr. 4 - 68930697 - 68951093 [-] |
GenBankmRNA |
BF132129.1 |
- |
3 |
204 |
|
Chr. 4 - 68930736 - 68951090 [-] |
GenBankmRNA |
DA043730.1 |
- |
3 |
478 |
|
Chr. 4 - 68943283 - 68951361 [-] |
View Full Product Details