Gene Transcripts
- Gene Symbol
- SNORA49
- Entrez Gene ID
- 677829
- Gene Name
- small nucleolar RNA, H/ACA box 49
- Gene Aliases
- ACA49
- Chromosome Location
- Chr. 12: 132031224 - 132031360 on Build GRCh38
- UniGene ID
- Hs.742405
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002979.2 |
|
1 |
29 |
|
View Full Product Details