Gene Transcripts
- Gene Symbol
- SNORA55
- Entrez Gene ID
- 677834
- Gene Name
- small nucleolar RNA, H/ACA box 55
- Gene Aliases
- ACA55
- Chromosome Location
- Chr. 1: 39567374 - 39567510 on Build GRCh38
- UniGene ID
- Hs.676660
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002983.1 |
|
1 |
127 |
|
GenBank mRNA |
AJ609480.1 |
- |
1 |
127 |
|
View Full Product Details