Gene Transcripts
- Gene Symbol
- SNORA60
- Entrez Gene ID
- 677837
- Gene Name
- small nucleolar RNA, H/ACA box 60
- Gene Aliases
- ACA60
- Chromosome Location
- Chr. 20: 38449369 - 38449504 on Build GRCh38
- UniGene ID
- Hs.16936
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002986.1 |
|
1 |
126 |
|
GenBank mRNA |
AJ609469.1 |
- |
1 |
126 |
|
AK091209.1 |
- |
1 |
1659 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BF807535.1 |
- |
1 |
180 |
|
Chr. 20 - 38449326 - 38449759 [+] |
GenBankmRNA |
AW411323.1 |
- |
1 |
110 |
|
Chr. 20 - 38449392 - 38450078 [+] |
GenBankmRNA |
BF308001.1 |
- |
1 |
102 |
|
Chr. 20 - 38449393 - 38450045 [+] |
GenBankmRNA |
BQ709000.1 |
- |
1 |
358 |
|
Chr. 20 - 38449137 - 38450027 [+] |
View Full Product Details