Gene Transcripts
- Gene Symbol
- SNORA71C
- Entrez Gene ID
- 677839
- Gene Name
- small nucleolar RNA, H/ACA box 71C
- Gene Aliases
- U71c
- Chromosome Location
- Chr. 20: 38429667 - 38429804 on Build GRCh38
- UniGene ID
- Hs.711516
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003017.2 |
|
1 |
125 |
|
View Full Product Details