Gene Transcripts

Gene Symbol
TTTY14
Entrez Gene ID
83869
Gene Name
testis-specific transcript, Y-linked 14 (non-protein coding)
Gene Aliases
CYorf14, NCRNA00137, NCRNA00185, PRO2834, TTY14
Chromosome Location
Chr. Y: 18872501 - 19077547 on Build GRCh38
UniGene ID
Hs.138453
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_125733.1
5 982
NR_125734.1
4 902
NR_125735.1
3 827
NR_125736.1
5 577
GenBank mRNA AF119903.1 - LIT2233
CYorf14
2 1344

View Full Product Details