Gene Transcripts

Gene Symbol
PLEKHM1P1
Entrez Gene ID
440456
Gene Name
pleckstrin homology and RUN domain containing M1 pseudogene 1
Gene Aliases
PLEKHM1P
Chromosome Location
Chr. 17: 64784841 - 64837184 on Build GRCh38
UniGene ID
Hs.646585
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024386.2 11 2634
GenBank mRNA AL833315.1 - 11 2604

View Full Product Details