Gene Transcripts

Gene Symbol
BAIAP2-AS1
Entrez Gene ID
440465
Gene Name
BAIAP2 antisense RNA 1 (head to head)
Gene Aliases
Chromosome Location
Chr. 17: 81029133 - 81034719 on Build GRCh38
UniGene ID
Hs.448889
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026857.1 2 1798
GenBank mRNA AK056555.1 - 2 1780
AK096609.1 - 1 705
AK124762.1 - 2 1839

View Full Product Details