Gene Transcripts
- Gene Symbol
- BAIAP2-AS1
- Entrez Gene ID
- 440465
- Gene Name
- BAIAP2 antisense RNA 1 (head to head)
- Gene Aliases
- Chromosome Location
- Chr. 17: 81029133 - 81034719 on Build GRCh38
- UniGene ID
- Hs.448889
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026857.1 |
|
2 |
3855 |
|
GenBank mRNA |
AK124762.1 |
- |
2 |
3896 |
|
BC110822.1 |
- |
1 |
1864 |
|
View Full Product Details