Gene Transcripts
- Gene Symbol
- GGT8P
- Entrez Gene ID
- 645367
- Gene Name
- gamma-glutamyltransferase 8 pseudogene
- Gene Aliases
- GGT1P1
- Chromosome Location
- Chr. 2: 91775342 - 91782127 on Build GRCh38
- UniGene ID
- Hs.650223
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003503.1 |
|
2 |
2461 |
|
GenBank mRNA |
AK055838.1 |
- |
2 |
2461 |
|
View Full Product Details