Gene Transcripts

Gene Symbol
FAAP100
Entrez Gene ID
80233
Gene Name
Fanconi anemia core complex associated protein 100
Gene Aliases
C17orf70
Chromosome Location
Chr. 17: 81539885 - 81552403 on Build GRCh38
UniGene ID
Hs.313905
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033338.1 2, 3 557
XM_017025162.1 XP_016880651.1 2, 3 382
GenBank mRNA BC008883.2 - 2, 3 530

View Full Product Details