Gene Transcripts
- Gene Symbol
- PPP2R2D
- Entrez Gene ID
- 55844
- Gene Name
- protein phosphatase 2 regulatory subunit Bdelta
- Gene Aliases
- MDS026
- Chromosome Location
- Chr. 10: 131900644 - 131971535 on Build GRCh38
- UniGene ID
- Hs.380372
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033191.2 |
|
|
736 |
|
View Full Product Details