Gene Transcripts
- Gene Symbol
- DIO3OS
- Entrez Gene ID
- 64150
- Gene Name
- DIO3 opposite strand/antisense RNA (head to head)
- Gene Aliases
- C14orf134, DIO3-AS1, DIO3-OS, NCRNA00041
- Chromosome Location
- Chr. 14: 101552223 - 101555676 on Build GRCh38
- UniGene ID
- Hs.525597
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002770.1 |
|
1 |
1960 |
|
GenBank mRNA |
AF469201.1 |
- |
2 |
979 |
|
AF469202.1 |
- |
1 |
1960 |
|
View Full Product Details