Gene Transcripts
- Gene Symbol
- GUSBP5
- Entrez Gene ID
- 441046
- Gene Name
- glucuronidase, beta pseudogene 5
- Gene Aliases
- GYPELOC441046
- Chromosome Location
- Chr. 4: 143559472 - 143561460 on Build GRCh38
- UniGene ID
- Hs.135705
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003675.2 |
|
1 |
1506 |
|
GenBank mRNA |
BC025996.2 |
- |
1 |
1506 |
|
View Full Product Details