Gene Transcripts
- Gene Symbol
- TTTY20
- Entrez Gene ID
- 252951
- Gene Name
- testis-specific transcript, Y-linked 20 (non-protein coding)
- Gene Aliases
- NCRNA00145
- Chromosome Location
- Chr. Y: 9329880 - 9334832 on Build GRCh38
- UniGene ID
- Hs.632292
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001546.1 |
|
2 |
143 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF527832.3 |
- |
2 |
143 |
|
Chr. Y - 9329880 - 9395788 [-] |
View Full Product Details