Gene Transcripts

Gene Symbol
LINC01548
Entrez Gene ID
728409
Gene Name
long intergenic non-protein coding RNA 1548
Gene Aliases
C21orf54
Chromosome Location
Chr. 21: 33165470 - 33170236 on Build GRCh38
UniGene ID
Hs.125234
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024102.1 3 357
GenBank mRNA BC105115.1 - 3 357
BC105117.1 - 3 357
BX116511.1 - 3 310


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AA400173.1 - 3 310 Chr. 21 - 33165535 - 33170189 [-]

View Full Product Details