Gene Transcripts
- Gene Symbol
- PCNAP1
- Entrez Gene ID
- 359806
- Gene Name
- proliferating cell nuclear antigen pseudogene 1
- Gene Aliases
- p1PCNA
- Chromosome Location
- Chr. 4: 99160593 - 99161647 on Build GRCh38
- UniGene ID
- Hs.537400
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028270.1 |
|
1 |
553 |
|
View Full Product Details