Gene Transcripts
- Gene Symbol
- ANTXRL
- Entrez Gene ID
- 195977
- Gene Name
- anthrax toxin receptor-like
- Gene Aliases
- Chromosome Location
- Chr. 10: 46286997 - 46330419 on Build GRCh38
- UniGene ID
- Hs.538515
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278688.2 |
NP_001265617.1
|
4, 5 |
749 |
|
XM_006717684.3 |
XP_006717747.1
|
4, 5 |
749 |
|
XM_011539430.2 |
XP_011537732.1
|
4, 5 |
749 |
|
XM_011539431.2 |
XP_011537733.1
|
3, 4 |
431 |
|
XM_011539433.2 |
XP_011537735.1
|
3, 4 |
538 |
|
XM_011539434.2 |
XP_011537736.1
|
3, 4 |
338 |
|
XM_011539437.2 |
XP_011537739.1
|
3, 4 |
857 |
|
XM_011539438.2 |
XP_011537740.1
|
3, 4 |
853 |
|
XM_011539439.2 |
XP_011537741.1
|
3, 4 |
1120 |
|
XM_011539440.2 |
XP_011537742.1
|
4, 5 |
917 |
|
XM_011539441.1 |
XP_011537743.1
|
4, 5 |
749 |
|
XM_011539442.1 |
XP_011537744.1
|
4, 5 |
749 |
|
XM_017015818.1 |
XP_016871307.1
|
3, 4 |
343 |
|
XM_017015819.1 |
XP_016871308.1
|
3, 4 |
1008 |
|
GenBank mRNA |
AK057316.1 |
- |
5, 6 |
450 |
|
View Full Product Details