Gene Transcripts

Gene Symbol
CECR5-AS1
Entrez Gene ID
100130717
Gene Name
CECR5 antisense RNA 1
Gene Aliases
CECR4, NCRNA00017
Chromosome Location
Chr. 22: 17159389 - 17165445 on Build GRCh38
UniGene ID
Hs.660066
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024482.1 2 305
NR_024483.1 2 591
GenBank mRNA AF307448.1 - 2 585


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000431923.1 - 2 581 Chr. 22 - 17159399 - 17165445 [+]
GenBankmRNA CK903337.1 - 2 321 Chr. 22 - 17159382 - 17165439 [+]
GENCODE ENST00000329743.3 - 1 310 Chr. 22 - 17159393 - 17159765 [+]

View Full Product Details