Gene Transcripts
- Gene Symbol
- HYMAI
- Entrez Gene ID
- 57061
- Gene Name
- hydatidiform mole associated and imprinted (non-protein coding)
- Gene Aliases
- NCRNA00020
- Chromosome Location
- Chr. 6: 144004916 - 144008730 on Build GRCh38
- UniGene ID
- Hs.657760
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002768.2 |
|
1 |
1740 |
|
GenBank mRNA |
AF241534.1 |
- |
1 |
1740 |
|
BC059359.1 |
- |
1 |
1261 |
|
View Full Product Details