Gene Transcripts
- Gene Symbol
- PRAC2
- Entrez Gene ID
- 360205
- Gene Name
- prostate cancer susceptibility candidate 2
- Gene Aliases
- C17orf93, HOXB-AS5, HOXB13-AS1, NCRNA00253
- Chromosome Location
- Chr. 17: 48720802 - 48724758 on Build GRCh38
- UniGene ID
- Hs.236557
- Species
- Homo sapiens
- Gene Symbol
- LOC102724665
- Entrez Gene ID
- 102724665
- Gene Name
- putative protein PRCA2
- Gene Aliases
- Chromosome Location
- Chr. 6: 92644258 - 92645174 on Build GRCh38
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB048981.1 |
- |
2 |
319 |
|
Chr. 17 - 48723209 - 48724729 [+] |
View Full Product Details