Gene Transcripts

Gene Symbol
OR6W1P
Entrez Gene ID
89883
Gene Name
olfactory receptor family 6 subfamily W member 1 pseudogene
Gene Aliases
OR6W1, sdolf
Chromosome Location
Chr. 7: 143062284 - 143063785 on Build GRCh38
UniGene ID
Hs.339818
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002140.1 1 1243
GenBank mRNA BC069382.1 - 1 834
BC074961.2 - 1 819
BC074962.2 - 1 819

View Full Product Details