Gene Transcripts
- Gene Symbol
- CASC15
- Entrez Gene ID
- 401237
- Gene Name
- cancer susceptibility candidate 15 (non-protein coding)
- Gene Aliases
- LINC00340
- Chromosome Location
- Chr. 6: 21666444 - 22194400 on Build GRCh38
- UniGene ID
- Hs.712707
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AU127831.2 |
- |
5, 6 |
496 |
|
Chr. 6 - 21668819 - 22020508 [+] |
GenBankmRNA |
DA507188.1 |
- |
5, 6 |
495 |
|
Chr. 6 - 21664234 - 22020408 [+] |
GenBankmRNA |
BM564069.1 |
- |
|
333 |
|
Chr. - - [-] |
GenBankmRNA |
CD674380.1 |
- |
|
426 |
|
Chr. 6 - 21885663 - 21979063 [+] |
GenBankmRNA |
CR739395.1 |
- |
3, 4 |
334 |
|
Chr. 6 - 21886230 - 22020405 [+] |
View Full Product Details