Gene Transcripts
- Gene Symbol
- FKSG29
- Entrez Gene ID
- 100131561
- Gene Name
- FKSG29
- Gene Aliases
- Chromosome Location
- Chr. 13: 99351420 - 99352027 on Build GRCh38
- UniGene ID
- Hs.721900
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024013.1 |
|
1 |
472 |
|
GenBank mRNA |
AY014273.1 |
- |
1 |
472 |
|
View Full Product Details