Gene Transcripts
- Gene Symbol
- RPL23P8
- Entrez Gene ID
- 222901
- Gene Name
- ribosomal protein L23 pseudogene 8
- Gene Aliases
- RPL23_4_790
- Chromosome Location
- Chr. 7: 20827298 - 20827820 on Build GRCh38
- UniGene ID
- Hs.722187
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026673.1 |
|
1 |
71 |
|
View Full Product Details