Gene Transcripts
- Gene Symbol
- HCG11
- Entrez Gene ID
- 493812
- Gene Name
- HLA complex group 11 (non-protein coding)
- Gene Aliases
- CTA-14H9.3, bK14H9.3
- Chromosome Location
- Chr. 6: 26521706 - 26527393 on Build GRCh38
- UniGene ID
- Hs.272939
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026790.1 |
|
1 |
1130 |
|
GenBank mRNA |
AK054919.1 |
- |
1 |
988 |
|
View Full Product Details