Gene Transcripts

Gene Symbol
SNHG6
Entrez Gene ID
641638
Gene Name
small nucleolar RNA host gene 6
Gene Aliases
HBII-276HG, NCRNA00058, U87HG
Chromosome Location
Chr. 8: 66921930 - 66925542 on Build GRCh38
UniGene ID
Hs.372680
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002599.1 4 399
GenBank mRNA AY264285.1 - 4 399


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI669107.1 - 4 453 Chr. 8 - 66921934 - 66925598 [-]
GenBankmRNA HY162808.1 - 4 398 Chr. 8 - 66921987 - 66925540 [-]
GenBankmRNA BM737762.1 - 4 386 Chr. 8 - 66921984 - 66925529 [-]
GenBankmRNA BQ924387.1 - 376 Chr. - - [-]
GenBankmRNA BI669733.1 - 4 404 Chr. 8 - 66921925 - 66925541 [-]
GenBankmRNA DA126867.1 - 4 398 Chr. 8 - 66921984 - 66925541 [-]
GenBankmRNA BM016905.1 - 4 383 Chr. 8 - 66921935 - 66925526 [-]
GenBankmRNA BY796444.2 - 4 374 Chr. 8 - 66921935 - 66925517 [-]
GenBankmRNA BM737568.1 - 397 Chr. - - [-]
GenBankmRNA BU193261.1 - 4 396 Chr. 8 - 66921934 - 66925530 [-]
GenBankmRNA BU167862.1 - 4 399 Chr. 8 - 66921925 - 66925534 [-]
GenBankmRNA BU193032.1 - 4 365 Chr. 8 - 66921934 - 66925508 [-]
GenBankmRNA BU195948.1 - 4 381 Chr. 8 - 66921968 - 66925524 [-]
GenBankmRNA CN257859.1 - 4 376 Chr. 8 - 66921938 - 66925519 [-]
GenBankmRNA AA633432.1 - 4 384 Chr. 8 - 66921984 - 66925524 [-]
GenBankmRNA BG034409.1 - 395 Chr. - - [-]
GenBankmRNA HY072040.1 - 4 397 Chr. 8 - 66921934 - 66925540 [-]
GenBankmRNA BM014460.1 - 4 383 Chr. 8 - 66921934 - 66925525 [-]
GenBankmRNA BM784351.1 - 4 398 Chr. 8 - 66921984 - 66925541 [-]
GenBankmRNA BF966290.2 - 4 402 Chr. 8 - 66921934 - 66925541 [-]
GenBankmRNA DB498305.1 - 4 449 Chr. 8 - 66921934 - 66925591 [-]
GenBankmRNA AW958356.1 - 383 Chr. - - [-]
GenBankmRNA BM768256.1 - 4 398 Chr. 8 - 66921968 - 66925541 [-]
GenBankmRNA AA429085.1 - 4 387 Chr. 8 - 66921958 - 66925527 [-]
GenBankmRNA BU596037.1 - 401 Chr. - - [-]
GenBankmRNA BQ440073.1 - 380 Chr. - - [-]
GenBankmRNA BM743937.1 - 4 397 Chr. 8 - 66921990 - 66925541 [-]
GenBankmRNA BI918140.1 - 384 Chr. - - [-]
GenBankmRNA CB963297.1 - 4 343 Chr. 8 - 66921984 - 66925481 [-]
GenBankmRNA BG032838.1 - 367 Chr. - - [-]
GenBankmRNA HY117390.1 - 4 449 Chr. 8 - 66921984 - 66925591 [-]
GenBankmRNA BG034121.1 - 4 388 Chr. 8 - 66921938 - 66925531 [-]
GenBankmRNA BM842017.1 - 4 399 Chr. 8 - 66921984 - 66925542 [-]
GenBankmRNA BM842145.1 - 4 399 Chr. 8 - 66921984 - 66925542 [-]
GenBankmRNA BE254660.1 - 361 Chr. - - [-]
GenBankmRNA BI550715.1 - 4 404 Chr. 8 - 66921934 - 66925542 [-]
GenBankmRNA BQ073523.1 - 376 Chr. - - [-]
GenBankmRNA BP425366.1 - 4 376 Chr. 8 - 66921938 - 66925511 [-]
GenBankmRNA HY066133.1 - 4 397 Chr. 8 - 66921948 - 66925540 [-]
GenBankmRNA BM741967.1 - 4 397 Chr. 8 - 66921991 - 66925541 [-]
GenBankmRNA CN482917.1 - 4 382 Chr. 8 - 66921934 - 66925525 [-]
GenBankmRNA AW630767.1 - 4 375 Chr. 8 - 66921971 - 66925518 [-]
GenBankmRNA HY112530.1 - 4 398 Chr. 8 - 66921948 - 66925540 [-]
GenBankmRNA HY075045.1 - 4 400 Chr. 8 - 66921984 - 66925540 [-]
GenBankmRNA DB569402.1 - 4 396 Chr. 8 - 66921936 - 66925539 [-]
GenBankmRNA CX752858.1 - 4 381 Chr. 8 - 66921934 - 66925524 [-]
GenBankmRNA CF129421.1 - 3 305 Chr. 8 - 66921934 - 66922723 [-]
GenBankmRNA DA301664.1 - 4 450 Chr. 8 - 66921910 - 66925593 [-]
GenBankmRNA BM016312.1 - 367 Chr. - - [-]
GenBankmRNA DA154192.1 - 4 398 Chr. 8 - 66921818 - 66925541 [-]
GenBankmRNA HY084735.1 - 4 397 Chr. 8 - 66921987 - 66925540 [-]
GenBankmRNA CB122847.1 - 4 370 Chr. 8 - 66921984 - 66925513 [-]
GenBankmRNA CD513777.1 - 4 410 Chr. 8 - 66921989 - 66925530 [-]
GenBankmRNA BE789325.1 - 4 351 Chr. 8 - 66921991 - 66925496 [-]
GenBankmRNA BU566037.1 - 355 Chr. - - [-]
GenBankmRNA CD513303.1 - 4 376 Chr. 8 - 66921984 - 66925519 [-]
GenBankmRNA F21431.1 - 2 370 Chr. 19 - 16889188 - 16890995 [+]
GenBankmRNA BM760648.1 - 4 399 Chr. 8 - 66921991 - 66925542 [-]
GenBankmRNA HY056578.1 - 4 397 Chr. 8 - 66921934 - 66925540 [-]
GenBankmRNA DA084810.1 - 4 399 Chr. 8 - 66921839 - 66925542 [-]
GenBankmRNA EL946282.1 - 4 390 Chr. 8 - 66921971 - 66925487 [-]
GenBankmRNA BM737712.1 - 4 384 Chr. 8 - 66921986 - 66925527 [-]
GenBankmRNA BU953849.1 - 4 425 Chr. 8 - 66921987 - 66925566 [-]
GenBankmRNA HY012009.1 - 4 397 Chr. 8 - 66921987 - 66925540 [-]
GenBankmRNA AA316590.1 - 4 370 Chr. 8 - 66921930 - 66925513 [-]
GenBankmRNA HY122223.1 - 4 396 Chr. 8 - 66921936 - 66925540 [-]
GenBankmRNA BQ229214.1 - 348 Chr. - - [-]
GenBankmRNA BM511306.1 - 4 354 Chr. 8 - 66921984 - 66925490 [-]

View Full Product Details