Gene Transcripts
- Gene Symbol
- GUCY1B2
- Entrez Gene ID
- 2974
- Gene Name
- guanylate cyclase 1 soluble subunit beta 2 (pseudogene)
- Gene Aliases
- GC-SB2
- Chromosome Location
- Chr. 13: 50994511 - 51066157 on Build GRCh38
- UniGene ID
- Hs.411573
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003923.2 |
|
1, 2 |
205 |
|
GenBank mRNA |
AF038499.2 |
- |
1, 2 |
54 |
|
AK296746.1 |
- |
2, 3 |
361 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA439280.1 |
- |
2, 3 |
361 |
|
Chr. 13 - 51034136 - 51080862 [-] |
GenBankmRNA |
AL530786.3 |
- |
2, 3 |
260 |
|
Chr. 13 - 51020564 - 51080805 [-] |
View Full Product Details