Gene Transcripts

A Gene Symbol is not available because the non-coding siRNA was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID. Non-coding siRNAs are designed to RefSeq (NR_) or to curated sequences from RNAdb represented by GenBank RNA or ESTdb accessions.


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CB204253.1 - 1 439 Chr. 2 - 174266431 - 174267044 [-]
GenBankmRNA AK084753.1 - 1 452 Chr. - - [-]
GenBankmRNA BB479526.2 - 1 442 Chr. 2 - 174266433 - 174267047 [-]
GenBankmRNA CB205747.1 - 1 431 Chr. 2 - 174266431 - 174267036 [-]

View Full Product Details