Gene Transcripts

Gene Symbol
Zim3
Entrez Gene ID
116811
Gene Name
zinc finger, imprinted 3
Gene Aliases
1700128I23Rik
Chromosome Location
Chr. 7: 6955685 - 6976662 on Build GRCm38
UniGene ID
Mm.381671
Species
Mus musculus
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_036631.2
6 929
GenBank mRNA AF365932.1 - LIT1822
Zim3
6 934

View Full Product Details