Gene Transcripts

Gene Symbol
SNRPN
Entrez Gene ID
6638
Gene Name
small nuclear ribonucleoprotein polypeptide N
Gene Aliases
HCERN3, PWCR, RT-LI, SM-D, SMN, SNRNP-N, SNURF-SNRPN, sm-N
Chromosome Location
Chr. 15: 24823637 - 24978723 on Build GRCh38
UniGene ID
Hs.555970
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF400487.1 - LIT2225
IC-SNURF-SNRPN 3 prime noncoding transcripts
5 658
BX648788.1 -
6 803


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BF948046.1 - 126 Chr. - - [-]
GenBankmRNA CN418825.1 - 1 530 Chr. 15 - 25089301 - 25089842 [+]
GenBankmRNA BE260901.1 - 384 Chr. - - [-]
GenBankmRNA CD654912.1 - 2 251 Chr. 15 - 25087576 - 25097361 [+]
GenBankmRNA DA120721.1 - 2 326 Chr. 15 - 25087491 - 25093852 [+]
GenBankmRNA CD643200.1 - 4 524 Chr. 15 - 25083496 - 25093923 [+]
GenBankmRNA DA737283.1 - 1 186 Chr. 15 - 25089645 - 25098745 [+]

View Full Product Details