Gene Transcripts

Gene Symbol
TRAM2
Entrez Gene ID
9697
Gene Name
translocation associated membrane protein 2
Gene Aliases
Chromosome Location
Chr. 6: 52497402 - 52577064 on Build GRCh38
UniGene ID
Hs.520182
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AY927571.1 - LIT3572
AY927571
1 21

View Full Product Details