Gene Transcripts

Gene Symbol
SNHG3
Entrez Gene ID
8420
Gene Name
small nucleolar RNA host gene 3
Gene Aliases
NCRNA00014, RNU17C, RNU17D, U17HG, U17HG-A, U17HG-AB
Chromosome Location
Chr. 1: 28505943 - 28510892 on Build GRCh38
UniGene ID
Hs.469723
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002909.2
3 1234
NR_036473.1
4 1342
GenBank mRNA AJ006835.1 - LIT1315
U17HG
3 1134
AK311577.1 -
1 868


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AL523116.3 - 3 1003 Chr. 1 - 28508127 - 28509940 [+]

View Full Product Details