Gene Transcripts

Gene Symbol
SNRPN
Entrez Gene ID
6638
Gene Name
small nuclear ribonucleoprotein polypeptide N
Gene Aliases
HCERN3, PWCR, RT-LI, SM-D, SMN, SNRNP-N, SNURF-SNRPN, sm-N
Chromosome Location
Chr. 15: 24823637 - 24978723 on Build GRCh38
UniGene ID
Hs.555970
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF400486.1 - LIT2209
IC-SNURF-SNRPN 3 prime noncoding transcripts
5 706
AL832758.1 -
3 307
AY362862.1 -
5 626
BC080646.1 -
3 316


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CN332353.1 - 1 38 Chr. 15 - 25061961 - 25073939 [+]
GenBankmRNA BX490842.1 - 4 597 Chr. 15 - 25056494 - 25062008 [+]

View Full Product Details