Gene Transcripts

Gene Symbol
C9orf3
Entrez Gene ID
84909
Gene Name
chromosome 9 open reading frame 3
Gene Aliases
AOPEP, AP-O, APO, C90RF3, ONPEP
Chromosome Location
Chr. 9: 94726604 - 95148264 on Build GRCh38
UniGene ID
Hs.434253
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017015230.1 XP_016870719.1 16 5086
XM_017015231.1 XP_016870720.1 14 4789
XM_017015237.1 XP_016870726.1 13 4622
GenBank mRNA AF043897.1 - 1 1821

View Full Product Details