Gene Transcripts
- Gene Symbol
- HBCBP
- Entrez Gene ID
- 100130083
- Gene Name
- HBcAg-binding protein
- Gene Aliases
- Chromosome Location
- Chr. 12: 56996710 - 56998440 on Build GRCh38
- UniGene ID
- Hs.539356
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF529371.1 |
- |
1 |
580 |
|
- Gene Symbol
- GPR182
- Entrez Gene ID
- 11318
- Gene Name
- G protein-coupled receptor 182
- Gene Aliases
- 7TMR, ADMR, AM-R, AMR, G10D, gamrh, hrhAMR
- Chromosome Location
- Chr. 12: 56993895 - 56998441 on Build GRCh38
- UniGene ID
- Hs.483909
- Species
- Homo sapiens
View Full Product Details