Gene Transcripts

Gene Symbol
CNNM2
Entrez Gene ID
54805
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 2
Gene Aliases
ACDP2, HOMG6, HOMGSMR
Chromosome Location
Chr. 10: 102918293 - 103087169 on Build GRCh38
UniGene ID
Hs.643509
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011539911.2 XP_011538213.1 3 3540
GenBank mRNA AK090785.1 - 1 697

View Full Product Details