Gene Transcripts

Gene Symbol
NHSL2
Entrez Gene ID
340527
Gene Name
NHS like 2
Gene Aliases
Chromosome Location
Chr. X: 71911073 - 72153286 on Build GRCh38
UniGene ID
Hs.397836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011530934.2 XP_011529236.1 8 16527
XM_017029473.1 XP_016884962.1 8 15497
XM_017029474.1 XP_016884963.1 8 11639
XM_017029475.1 XP_016884964.1 4 14882
XM_017029476.1 XP_016884965.1 4 10999
XM_017029477.1 XP_016884966.1 4 15913
GenBank mRNA AK056217.1 - 1 749


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA682621.1 - 1 295 Chr. X - 72151337 - 72151903 [+]
GenBankmRNA BE620108.1 - 1 209 Chr. X - 72151422 - 72152229 [+]
GenBankmRNA CX752026.1 - 1 318 Chr. X - 72151323 - 72152030 [+]

View Full Product Details